Assay Details
Target Gene Details
Entrez Gene ID: | 13602 |
Gene Name: | SPARC-like 1 |
Gene Aliases: |
Ecm2, Sc1, hevin, mast9 |
Location: |
Chr.5:104079108-104114088 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sparcl1 | NM_010097.4 | NP_034227.3 | ||
| AK134608.1 | BAE22205.1 | |||
| AK158725.1 | BAE34628.1 | |||
| BC003759.1 | AAH03759.1 | |||
| CD742522.1 | ||||
| U64827.1 | AAB08451.1 | |||
| U77330.1 | AAC53172.1 |
More Information
Set Membership: |
|

Genomic Map