Assay Details
Target Gene Details
Entrez Gene ID: | 231642 |
Gene Name: | alkB, alkylation repair homolog 2 (E. coli) |
Gene Aliases: |
9530023G02, AU016977, Abh2, mABH2 |
Location: |
Chr.5:114123926-114130546 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Alkbh2 |
Target Gene Details
Entrez Gene ID: | 22256 |
Gene Name: | uracil DNA glycosylase |
Gene Aliases: |
UNG1, UNG2 |
Location: |
Chr.5:114130360-114139321 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ung | NM_001040691.1 | 1 | 99 | NP_001035781.1 |
| AK146178.1 | 1 | 99 | BAE26956.1 | |
| BC011039.1 | 1 | 156 | AAH11039.1 | |
| Y08975.1 | 1 | 99 | CAA70168.1 |
More Information
Set Membership: |
|

Genomic Map