Assay Details
Target Gene Details
Entrez Gene ID: | 22256 |
Gene Name: | uracil DNA glycosylase |
Gene Aliases: |
UNG1, UNG2 |
Location: |
Chr.5:114130360-114139321 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ung | NM_001040691.1 | 6 | 753 | NP_001035781.1 |
| NM_011677.2 | 5 | 725 | NP_035807.2 | |
| AK146178.1 | 6 | 753 | BAE26956.1 | |
| AK168709.1 | 5 | 675 | BAE40551.1 | |
| AK168785.1 | 5 | 675 | BAE40619.1 | |
| BC011039.1 | 6 | 810 | AAH11039.1 | |
| BC052853.1 | 5 | 669 | AAH52853.1 | |
| U55041.1 | 5 | 679 | AAC53197.1 | |
| X99018.1 | 5 | 681 | CAA67489.1 | |
| Y08975.1 | 6 | 753 | CAA70168.1 |
More Information
Set Membership: |
|

Genomic Map