Assay Details
Target Gene Details
Entrez Gene ID: | 30956 |
Gene Name: | aminoadipate-semialdehyde synthase |
Gene Aliases: |
LKR, LKR/SDH, LOR, LOR/SDH, Lorsdh, SDH |
Location: |
Chr.6:23072173-23132986 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aass | NM_013930.4 | NP_038958.2 | ||
| AJ224761.1 | CAA12114.1 | |||
| AK044323.1 | ||||
| AK098141.1 | ||||
| AK136216.1 | BAE22882.1 | |||
| AK149400.1 | BAE28852.1 | |||
| BC005420.1 | AAH05420.1 |
More Information
Set Membership: |
|

Genomic Map