Assay Details
Target Gene Details
Entrez Gene ID: | 319653 |
Gene Name: | solute carrier family 25, member 40 |
Gene Aliases: |
B230315F11Rik |
Location: |
Chr.5:8422838-8454840 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a40 | NM_001289595.1 | NP_001276524.1 | ||
| NM_001289596.1 | NP_001276525.1 | |||
| NM_178766.5 | NP_848881.2 | |||
| AK045863.1 | BAC32513.1 | |||
| AK050815.1 | ||||
| AK079538.1 | ||||
| AK084797.1 | BAC39281.1 | |||
| AK088444.1 | BAC40356.1 | |||
| AK136770.1 | BAE23124.1 | |||
| AK163037.1 | ||||
| BC083103.1 | AAH83103.1 |

Genomic Map