Assay Details
Target Gene Details
Entrez Gene ID: | 80979 |
Gene Name: | solute carrier family 26, member 5 |
Gene Aliases: |
Pres, prestin |
Location: |
Chr.5:21809001-21865604 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc26a5 | NM_001289787.1 | 2 | 215 | NP_001276716.1 |
| NM_001289788.1 | 2 | 215 | NP_001276717.1 | |
| NM_030727.5 | 3 | 290 | NP_109652.3 | |
| AF529192.1 | 3 | 290 | AAO59381.1 | |
| AY024359.2 | AAG59999.2 | |||
| BC108986.1 | 2 | 207 | AAI08987.1 | |
| BC108987.1 | 2 | 207 | AAI08988.1 |
More Information
Set Membership: |
|

Genomic Map