Assay Details
Target Gene Details
Entrez Gene ID: | 69852 |
Gene Name: | transcription factor 23 |
Gene Aliases: |
2010002O16Rik, Out, bHLHa24 |
Location: |
Chr.5:30968601-30988065 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tcf23 | NM_053085.2 | 2 | 403 | NP_444315.1 |
| AF142405.1 | 2 | 403 | AAF31168.1 | |
| AK030277.1 | 2 | 419 | BAC26873.1 | |
| AK030297.1 | 2 | 409 | BAC26885.1 | |
| BC119346.1 | 2 | 374 | ||
| BC120609.1 | 2 | 374 |
More Information
Set Membership: |
|

Genomic Map