Assay Details
Target Gene Details
Entrez Gene ID: | 66993 |
Gene Name: | SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily d, member 3 |
Gene Aliases: |
1500001J14Rik, 2210409C08Rik, BAF60C |
Location: |
Chr.5:24592622-24624456 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smarcd3 | NM_025891.3 | NP_080167.3 | ||
| AK005094.1 | BAB23813.1 | |||
| BC013122.1 | AAH13122.2 | |||
| BC060525.1 | AAH60525.1 |
More Information
Set Membership: |
|

Genomic Map