Assay Details
Target Gene Details
Entrez Gene ID: | 70118 |
Gene Name: | SRR1 domain containing |
Gene Aliases: |
2810002G02Rik, Srr1 |
Location: |
Chr.5:112337391-112343115 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Srrd | NM_027323.2 | NP_081599.2 | ||
| AK012640.1 | BAB28374.1 | |||
| BC030678.1 | AAH30678.1 |

Genomic Map