Assay Details
Target Gene Details
Entrez Gene ID: | 70291 |
Gene Name: | makorin, ring finger protein 2, opposite strand |
Gene Aliases: |
2510049J12Rik |
Location: |
Chr.6:115583547-115592576 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mkrn2os | NM_001101431.1 | 3 | 443 | NP_001094901.1 |
| AA277646.1 | ||||
| AA833313.1 | 3 | 345 | ||
| AK011120.1 | 2 | 366 | ||
| AK028760.1 | 2 | 316 |

Genomic Map