Assay Details
Target Gene Details
Entrez Gene ID: | 11810 |
Gene Name: | apolipoprotein B mRNA editing enzyme, catalytic polypeptide 1 |
Gene Aliases: |
Cdar1 |
Location: |
Chr.6:122577792-122603041 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Apobec1 | NM_001134391.1 | 4 | 463 | NP_001127863.1 |
| NM_031159.3 | 4 | 519 | NP_112436.1 | |
| AK146427.1 | 4 | 395 | BAE27161.1 | |
| AK151799.1 | 4 | 391 | BAE30699.1 | |
| AK165209.1 | 4 | 402 | BAE38079.1 | |
| AK166412.1 | 4 | 395 | BAE38760.1 | |
| BC003792.1 | 4 | 407 | AAH03792.1 | |
| U22262.1 | 4 | 519 | AAC52212.1 | |
| U22263.1 | 4 | 463 | AAC52213.1 | |
| U22264.1 | 1 | 219 | AAC52214.1 |
More Information
Set Membership: |
|

Genomic Map