Assay Details
Target Gene Details
Entrez Gene ID: | 66144 |
Gene Name: | ATPase, H+ transporting, lysosomal V1 subunit F |
Gene Aliases: |
1110004G16Rik |
Location: |
Chr.6:29467783-29470509 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v1f | NM_025381.2 | NP_079657.1 | ||
| AK003419.1 | BAB22780.1 | |||
| AK007327.1 | BAB24962.1 | |||
| AK152153.1 | BAE30988.1 | |||
| AK152929.1 | BAE31604.1 | |||
| AK152959.1 | BAE31622.1 | |||
| BC016553.1 | AAH16553.1 |
More Information
Set Membership: |
|

Genomic Map