Assay Details
Target Gene Details
Entrez Gene ID: | 11993 |
Gene Name: | ancient ubiquitous protein 1 |
Gene Aliases: |
AA589454 |
Location: |
Chr.6:83054521-83057682 on Build GRCm38 |
Assay Gene Location: | Within Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aup1 | NM_001301649.1 | NP_001288578.1 | ||
| NM_007517.4 | NP_031543.2 | |||
| NR_125891.1 | ||||
| AK144497.1 | 1 | 1291 | ||
| AK154707.1 | BAE32776.1 | |||
| AK158035.1 | BAE34327.1 | |||
| AK167335.1 | ||||
| AK171011.1 | BAE42182.1 | |||
| AK171184.1 | ||||
| AK171202.1 | ||||
| AK171249.1 | ||||
| AK171541.1 | BAE42515.1 | |||
| AK171621.1 | ||||
| AK187544.1 | ||||
| AK210654.1 | ||||
| BC016485.1 | AAH16485.1 | |||
| U41736.1 | AAC52839.1 |
More Information
Set Membership: |
|

Genomic Map