Assay Details
Target Gene Details
Entrez Gene ID: | 13835 |
Gene Name: | Eph receptor A1 |
Gene Aliases: |
5730453L17Rik, AL033318, Eph, Esk |
Location: |
Chr.6:42358487-42373310 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Epha1 | NM_023580.4 | NP_076069.2 | ||
| AF131197.1 | AAG12206.1 | |||
| AK028478.1 | BAC25971.1 | |||
| BC071215.1 | AAH71215.1 | |||
| U18084.1 | AAC52384.1 |
More Information
Set Membership: |
|

Genomic Map