Assay Details
Target Gene Details
Entrez Gene ID: | 232371 |
Gene Name: | complement component 1, r subcomponent-like |
Gene Aliases: |
C1r-LP, C1rl1, C1rlp |
Location: |
Chr.6:124493113-124510643 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1rl | NM_181344.5 | NP_851989.3 | ||
| AF456428.1 | AAP41417.1 | |||
| AK038659.1 | ||||
| AK133495.1 | BAE21686.1 | |||
| AK134207.1 | BAE22052.1 | |||
| AK162497.1 | ||||
| BC070472.1 | AAH70472.1 |
More Information
Set Membership: |
|

Genomic Map