Assay Details
Target Gene Details
Entrez Gene ID: | 232371 |
Gene Name: | complement component 1, r subcomponent-like |
Gene Aliases: |
C1r-LP, C1rl1, C1rlp |
Location: |
Chr.6:124493113-124510643 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C1rl | NM_181344.5 | 3 | 477 | NP_851989.3 |
| AF456428.1 | 2 | 361 | AAP41417.1 | |
| AK038659.1 | 3 | 442 | ||
| AK133495.1 | 3 | 456 | BAE21686.1 | |
| AK134207.1 | 3 | 478 | BAE22052.1 | |
| AK162497.1 | 3 | 432 | ||
| BC070472.1 | 3 | 440 | AAH70472.1 |
More Information
Set Membership: |
|

Genomic Map