Assay Details
Target Gene Details
Entrez Gene ID: | 12525 |
Gene Name: | CD8 antigen, alpha chain |
Gene Aliases: |
BB154331, Ly-2, Ly-35, Ly-B, Lyt-2 |
Location: |
Chr.6:71352772-71379171 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cd8a | NM_001081110.2 | NP_001074579.1 | ||
| NM_009857.1 | NP_033987.1 | |||
| AK017889.1 | ||||
| AK037467.1 | BAC29812.1 | |||
| AK088128.1 | BAC40163.1 | |||
| AK088214.1 | BAC40215.1 | |||
| AK088414.1 | BAC40342.1 | |||
| AK088447.1 | ||||
| AK137937.1 | BAE23516.1 | |||
| BC030679.1 | AAH30679.1 | |||
| M12052.1 | AAA39478.1 | |||
| M12825.1 | AAA39476.1 | |||
| M16981.1 | AAA39477.1 | |||
| U34881.1 | AAA92533.1 |

Genomic Map