Assay Details
Target Gene Details
Entrez Gene ID: | 13848 |
Gene Name: | Eph receptor B6 |
Gene Aliases: |
Cekl, Mep |
Location: |
Chr.6:41605040-41620507 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ephb6 | NM_001146351.1 | NP_001139823.1 | ||
| NM_007680.4 | NP_031706.3 | |||
| AK076442.1 | ||||
| AK087423.1 | BAC39868.1 | |||
| BC031924.1 | AAH31924.1 | |||
| L77867.1 | AAB51430.1 |
More Information
Set Membership: |
|

Genomic Map