Assay Details
Target Gene Details
Entrez Gene ID: | 17837 |
Gene Name: | murinoglobulin 2 |
Gene Aliases: |
Cpamd8 |
Location: |
Chr.6:122006719-122085968 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 20 - Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mug2 | NM_008646.3 | NP_032672.2 | ||
| M65238.1 | AAA73041.1 |
More Information
Set Membership: |
|

Genomic Map