Assay Details
Target Gene Details
Entrez Gene ID: | 11517 |
Gene Name: | adenylate cyclase activating polypeptide 1 receptor 1 |
Gene Aliases: |
2900024I10Rik, AI846590, PAC1, PAC1R, PACAP1-R |
Location: |
Chr.6:55451501-55501455 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adcyap1r1 | NM_001025372.2 | 3 | 570 | NP_001020543.1 |
| NM_007407.4 | 3 | 570 | NP_031433.3 | |
| AK042820.1 | 3 | 562 | BAC31372.1 | |
| AK047304.1 | 2 | 73 | BAC33020.1 | |
| AK081121.1 | 2 | 73 | BAC38140.1 | |
| AK083108.1 | 3 | 568 | ||
| AK147291.1 | 3 | 521 | BAE27825.1 | |
| AK147609.1 | 3 | 311 | BAE28023.1 | |
| BC067039.1 | 3 | 503 | AAH67039.1 | |
| D82935.1 | 3 | 253 | BAA11639.1 |
More Information
Set Membership: |
|

Genomic Map