Assay Details
Target Gene Details
Entrez Gene ID: | 20893 |
Gene Name: | basic helix-loop-helix family, member e40 |
Gene Aliases: |
Bhlhb2, C130042M06Rik, CR8, Clast5, Dec1, Sharp2, Stra13, Stra14 |
Location: |
Chr.6:108660629-108666925 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bhlhe40 | NM_011498.4 | NP_035628.1 | ||
| AF010305.1 | AAB64228.1 | |||
| AF364051.1 | AAK50859.1 | |||
| AK048242.1 | BAC33281.1 | |||
| AK090032.1 | BAC41056.1 | |||
| AK154935.1 | BAE32936.1 | |||
| BC010720.1 | AAH10720.1 | |||
| Y07836.1 | CAA69169.1 |
More Information
Set Membership: |
|

Genomic Map