Assay Details
Target Gene Details
Entrez Gene ID: | 320405 |
Gene Name: | Ca2+-dependent activator protein for secretion 2 |
Gene Aliases: |
A230044C21Rik, Caps-2, Caps2, Cpd2 |
Location: |
Chr.6:23262773-23839421 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cadps2 | NM_001252105.1 | NP_001239034.1 | ||
| NM_001252106.1 | NP_001239035.1 | |||
| NM_001252107.1 | NP_001239036.1 | |||
| NM_001252108.1 | NP_001239037.1 | |||
| NM_001252110.1 | NP_001239039.1 | |||
| NM_153163.4 | NP_694803.3 | |||
| AB098623.1 | BAD05017.1 | |||
| AB291947.1 | ||||
| AB291948.1 | ||||
| AB291949.1 | ||||
| AB291950.1 | ||||
| AF000969.2 | AAB58720.2 | |||
| AK038568.1 | BAC30047.1 | |||
| AK046596.1 | BAC32801.1 | |||
| AY072800.1 | AAL67937.1 | |||
| BC047394.1 | AAH47394.1 | |||
| BC055462.1 | AAH55462.1 | |||
| BC059274.1 | AAH59274.1 | |||
| BC080854.1 | AAH80854.1 |
More Information
Set Membership: |
|

Genomic Map