Assay Details
Target Gene Details
Entrez Gene ID: | 13448 |
Gene Name: | docking protein 1 |
Gene Aliases: |
AW557123, p62DOK |
Location: |
Chr.6:83030932-83033471 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dok1 | NM_001291799.1 | NP_001278728.1 | ||
| NM_010070.4 | NP_034200.4 | |||
| AA790529.1 | ||||
| AK136382.1 | BAE22956.1 | |||
| AK169871.1 | ||||
| BC013066.1 | AAH13066.1 | |||
| CJ053718.1 | ||||
| U78818.2 | AAB48827.2 |

Genomic Map