Assay Details
Target Gene Details
Entrez Gene ID: | 76252 |
Gene Name: | ATPase, H+ transporting, lysosomal V0 subunit E2 |
Gene Aliases: |
0610006O14Rik, NM9.2 |
Location: |
Chr.6:48537569-48541801 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v0e2 | NM_133764.3 | NP_598525.1 | ||
| AK002260.1 | ||||
| AK077313.1 | ||||
| BC010790.1 | AAH10790.1 | |||
| BI134693.1 | ||||
| BY296901.1 |
More Information
Set Membership: |
|

Genomic Map