Assay Details
Target Gene Details
Entrez Gene ID: | 77042 |
Gene Name: | hyaluronoglucosaminidase 4 |
Gene Aliases: |
4632428M18Rik, CSHY |
Location: |
Chr.6:24748367-24766519 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hyal4 | NM_029848.1 | NP_084124.1 | ||
| AB524072.1 | ||||
| BC125402.1 | ||||
| BC132096.1 |
More Information
Set Membership: |
|

Genomic Map