Assay Details
Target Gene Details
Entrez Gene ID: | 28019 |
Gene Name: | inhibitor of growth family, member 4 |
Gene Aliases: |
D6Wsu147e, D6Xrf92, p29ING4 |
Location: |
Chr.6:125039848-125049264 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ing4 | NM_133345.2 | NP_579923.1 | ||
| AK002821.1 | BAC25009.1 | |||
| AK009267.1 | BAB26183.1 | |||
| AK031633.1 | BAC27489.1 | |||
| AK046126.1 | ||||
| AK050522.1 | BAC34304.1 | |||
| AY035880.1 | AAK63168.1 | |||
| AY035881.1 | AAK63169.1 | |||
| AY036107.1 | AAK64509.1 | |||
| BC009127.1 | AAH09127.1 |
More Information
Set Membership: |
|

Genomic Map