Assay Details
Target Gene Details
Entrez Gene ID: | 231997 |
Gene Name: | FK506 binding protein 14 |
Gene Aliases: |
BC029109, FKBP-14, FKBP22 |
Location: |
Chr.6:54577605-54593698 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fkbp14 | NM_153573.1 | NP_705801.1 | ||
| AK030620.1 | ||||
| AK044876.1 | ||||
| AK081692.1 | BAC38295.1 | |||
| BC029109.1 | AAH29109.1 | |||
| BC034313.1 |
More Information
Set Membership: |
|

Genomic Map