Assay Details
Target Gene Details
Entrez Gene ID: | 18575 |
Gene Name: | phosphodiesterase 1C |
Gene Aliases: |
- |
Location: |
Chr.6:56069798-56652516 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde1c | NM_001025568.2 | NP_001020739.1 | ||
| NM_001159952.1 | NP_001153424.1 | |||
| NM_001159953.1 | NP_001153425.1 | |||
| NM_001159955.1 | NP_001153427.1 | |||
| NM_001159956.1 | NP_001153428.1 | |||
| NM_001159957.1 | NP_001153429.1 | |||
| NM_001159960.1 | NP_001153432.1 | |||
| NM_011054.4 | 1 | 126 | NP_035184.1 | |
| AK014887.1 | BAB29604.1 | |||
| AK015375.1 | ||||
| AK017183.1 | ||||
| AK029531.1 | BAC26499.1 | |||
| AK030423.1 | BAC26956.1 | |||
| AK050499.1 | ||||
| AK077123.1 | BAC36626.1 | |||
| AK082180.1 | BAC38431.1 | |||
| AK133243.1 | ||||
| AK139923.1 | BAE24184.1 | |||
| BB614593.1 | ||||
| BB654130.1 | ||||
| L76946.1 | 1 | 127 | AAC37703.1 |
More Information
Set Membership: |
|

Genomic Map