Assay Details
Target Gene Details
Entrez Gene ID: | 78600 |
Gene Name: | phosphodiesterase 6H, cGMP-specific, cone, gamma |
Gene Aliases: |
A930033D18Rik, PDEgamma |
Location: |
Chr.6:136952221-136963485 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde6h | NM_023898.4 | NP_076387.2 | ||
| AF189146.1 | AAG17113.1 | |||
| AF190929.1 | AAG17170.1 | |||
| AK020922.1 | BAB32255.1 | |||
| AK080696.1 | ||||
| BC115671.1 | ||||
| BC115672.1 |
More Information
Set Membership: |
|

Genomic Map