Assay Details
Target Gene Details
Entrez Gene ID: | 66283 |
Gene Name: | gastrokine 1 |
Gene Aliases: |
2200002K21Rik, AMP-18, BRICD1, Fov |
Location: |
Chr.6:87345653-87350915 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gkn1 | NM_025466.1 | NP_079742.1 | ||
| AK008622.1 | BAB25784.1 | |||
| AK008640.1 | ||||
| AK008641.1 | BAB25801.1 | |||
| AK008647.1 | BAB25805.1 | |||
| AK008722.1 | BAB25856.1 | |||
| AK008745.1 | BAB25872.1 | |||
| AK008933.1 | BAB25975.1 | |||
| AK008956.1 | ||||
| AK008990.1 | BAB26010.1 | |||
| AK009145.1 | BAB26103.1 | |||
| AK019050.1 | BAB31525.3 | |||
| AK028136.1 | BAE20436.1 | |||
| AY456960.1 | AAR21210.1 | |||
| BC099446.1 | AAH99446.1 | |||
| BC132540.1 | ||||
| BC132542.1 |

Genomic Map