Assay Details
Target Gene Details
Entrez Gene ID: | 16950 |
Gene Name: | lysyl oxidase-like 3 |
Gene Aliases: |
Lor2, Loxl2 |
Location: |
Chr.6:83034224-83052564 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Loxl3 | NM_013586.4 | 8 | 1473 | NP_038614.2 |
| AF053368.1 | 8 | 1504 | AAC83205.1 | |
| AK030548.1 | 8 | 1474 | BAC27016.1 | |
| AK145189.1 | BAE26282.1 | |||
| BC011298.1 | 8 | 1467 | AAH11298.1 |
More Information
Set Membership: |
|

Genomic Map