Assay Details
Target Gene Details
Entrez Gene ID: | 171095 |
Gene Name: | interleukin 17 receptor C |
Gene Aliases: |
1110025H02Rik, Gm19850, IL-17RC, IL17-RC, IL17-RL, Il17rl |
Location: |
Chr.6:113471455-113483163 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Il17rc | NM_134159.4 | 1 | 135 | NP_598920.3 |
| NM_178942.1 | 1 | 135 | NP_849273.1 | |
| AF458066.1 | AAM77570.1 | |||
| BC004759.1 | 1 | 83 | AAH04759.1 | |
| BY018830.1 | 1 | 136 |
More Information
Set Membership: |
|

Genomic Map