Assay Details
Target Gene Details
Entrez Gene ID: | 67602 |
Gene Name: | NECAP endocytosis associated 1 |
Gene Aliases: |
1200016B17Rik, AI747569, mFLJ00061 |
Location: |
Chr.6:122874557-122888941 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Necap1 | NM_026267.2 | NP_080543.2 | ||
| AK004805.1 | BAB23577.2 | |||
| AK005263.1 | BAB23915.2 | |||
| AK081598.1 | BAC38266.1 | |||
| AK168500.1 | BAE40385.1 | |||
| AK220236.1 | BAD90161.1 | |||
| BC011466.1 | ||||
| BC119133.1 | ||||
| BC120841.1 |
More Information
Set Membership: |
|

Genomic Map