Assay Details
Target Gene Details
Entrez Gene ID: | 76522 |
Gene Name: | LSM8 homolog, U6 small nuclear RNA associated (S. cerevisiae) |
Gene Aliases: |
2010003I05Rik, AW214405, Naa38 |
Location: |
Chr.6:18848635-18854052 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lsm8 | NM_133939.1 | NP_598700.1 | ||
| AK008067.1 | ||||
| AK088664.1 | BAC40488.1 | |||
| BC019458.1 | AAH19458.1 |
More Information
Set Membership: |
|

Genomic Map