Assay Details
Target Gene Details
Entrez Gene ID: | 66223 |
Gene Name: | mitochondrial ribosomal protein L35 |
Gene Aliases: |
1110066C01Rik |
Location: |
Chr.6:71812997-71823784 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl35 | NM_025430.3 | NP_079706.1 | ||
| AK004385.1 | BAB23282.1 | |||
| AK010223.1 | BAB26778.1 | |||
| AK010369.1 | BAB26887.1 | |||
| AK012807.1 | BAB28485.1 | |||
| AK039187.1 | BAC30269.1 | |||
| BB615775.1 | ||||
| BC028750.1 | AAH28750.1 |
More Information
Set Membership: |
|

Genomic Map