Assay Details
Target Gene Details
Entrez Gene ID: | 18575 |
Gene Name: | phosphodiesterase 1C |
Gene Aliases: |
- |
Location: |
Chr.6:56069798-56652516 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 21 - Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pde1c | NM_001159953.1 | NP_001153425.1 | ||
| AK030423.1 | BAC26956.1 | |||
| AK133243.1 | ||||
| L76947.1 | AAC37702.1 |
More Information
Set Membership: |
|

Genomic Map