Assay Details
Target Gene Details
Entrez Gene ID: | 64704 |
Gene Name: | HtrA serine peptidase 2 |
Gene Aliases: |
AI481710, Omi, Prss25, mnd2 |
Location: |
Chr.6:83051266-83054571 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Htra2 | NM_019752.3 | NP_062726.3 | ||
| AF164513.1 | AAF89534.1 | |||
| AF175324.1 | AAD50499.1 | |||
| AK146338.1 | BAE27092.1 | |||
| AK159188.1 | BAE34885.1 | |||
| AK176936.1 | ||||
| AK178298.1 | ||||
| AK179655.1 | ||||
| AK182973.1 | ||||
| AK186135.1 | ||||
| AK188221.1 | ||||
| AK215600.1 | ||||
| BC049880.1 | AAH49880.1 |
More Information
Set Membership: |
|

Genomic Map