Assay Details
Target Gene Details
Entrez Gene ID: | 71395 |
Gene Name: | RIKEN cDNA 5430419D17 gene |
Gene Aliases: |
- |
Location: |
Chr.7:131174402-131282194 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 14 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 5430419D17Rik | NM_175166.3 | NP_780375.2 | ||
| AK017319.1 | 7 | 847 | ||
| AK035662.1 | BAC29143.1 |
More Information
Set Membership: |
|

Genomic Map