Assay Details
Target Gene Details
Entrez Gene ID: | 56463 |
Gene Name: | staphylococcal nuclease and tudor domain containing 1 |
Gene Aliases: |
AL033314, Tudor-SN |
Location: |
Chr.6:28480315-28935162 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snd1 | NM_019776.2 | 9 | 1213 | NP_062750.2 |
| AB021491.1 | 9 | 1170 | BAA84944.1 | |
| AK034914.1 | 8 | 1082 | ||
| AK088028.1 | 9 | 1172 | BAC40105.1 | |
| AK133837.1 | 9 | 1219 | BAE21874.1 | |
| AK159837.1 | 9 | 1178 | BAE35415.1 | |
| AK161591.1 | 9 | 1167 | BAE36479.1 | |
| AK162432.1 | 9 | 1154 | BAE36913.1 | |
| AK167576.1 | 9 | 1203 | BAE39639.1 | |
| AK185603.1 | 2 | 81 | ||
| AK190012.1 | 2 | 81 | ||
| AK196687.1 | 2 | 81 | ||
| AK200045.1 | 2 | 81 | ||
| AK216953.1 | 2 | 81 | ||
| BC007126.1 | 9 | 1160 | AAH07126.3 |

Genomic Map