Assay Details
Target Gene Details
Entrez Gene ID: | 66367 |
Gene Name: | RIKEN cDNA 2310022A10 gene |
Gene Aliases: |
- |
Location: |
Chr.7:27553260-27582099 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 2310022A10Rik | NM_175107.5 | NP_780316.3 | ||
| BB853451.1 |
Target Gene Details
Entrez Gene ID: | 18807 |
Gene Name: | phospholipase D family, member 3 |
Gene Aliases: |
Sam-9 |
Location: |
Chr.7:27531645-27553213 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pld3 |

Genomic Map