Assay Details
Target Gene Details
Entrez Gene ID: | 20690 |
Gene Name: | sperm adhesion molecule 1 |
Gene Aliases: |
AV039194, Ph-20 |
Location: |
Chr.6:24791188-24801048 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spam1 | NM_001079875.2 | NP_001073344.1 | ||
| NM_009241.3 | NP_033267.2 | |||
| AK005638.1 | BAB24161.1 | |||
| AY228460.1 | AAP49832.1 | |||
| AY920282.1 | AAX13989.1 | |||
| AY920283.1 | AAX13990.1 | |||
| U33958.1 | AAA76603.1 |
More Information
Set Membership: |
|

Genomic Map