Assay Details
Target Gene Details
Entrez Gene ID: | 50799 |
Gene Name: | solute carrier family 25 (mitochondrial carrier, adenine nucleotide translocator), member 13 |
Gene Aliases: |
AI785475, Ctrn |
Location: |
Chr.6:6041218-6217233 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 15 - Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc25a13 | NM_001177572.1 | NP_001171043.1 | ||
| NM_015829.3 | NP_056644.1 | |||
| AF164632.1 | AAF21426.1 | |||
| AK002829.1 | BAB22390.1 | |||
| AK012670.1 | BAB28397.1 | |||
| AK029020.1 | BAC26247.1 | |||
| AK150496.1 | ||||
| AK151180.1 | ||||
| AK152237.1 | ||||
| AK199748.1 | ||||
| BC016571.1 | AAH16571.1 |
More Information
Set Membership: |
|

Genomic Map