Assay Details
Target Gene Details
Entrez Gene ID: | 140491 |
Gene Name: | protein phosphatase 1, regulatory (inhibitor) subunit 3A |
Gene Aliases: |
GM, RG1, RGL |
Location: |
Chr.6:14713822-14755274 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r3a | NM_080464.2 | NP_536712.2 | ||
| AK084719.1 | BAC39262.2 | |||
| BC109007.1 | AAI09008.1 |
More Information
Set Membership: |
|

Genomic Map