Assay Details
Target Gene Details
Entrez Gene ID: | 232491 |
Gene Name: | pyridine nucleotide-disulphide oxidoreductase domain 1 |
Gene Aliases: |
- |
Location: |
Chr.6:142345625-142362630 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pyroxd1 | NM_183165.3 | NP_898988.2 | ||
| AK165677.1 | BAE38333.1 | |||
| BC027061.1 | AAH27061.1 |
More Information
Set Membership: |
|

Genomic Map