Assay Details
Target Gene Details
Entrez Gene ID: | 58887 |
Gene Name: | replication initiator 1 |
Gene Aliases: |
AI425994, Ap4, E430037F08Rik, Zfp-464, Zfp464 |
Location: |
Chr.6:48593883-48599082 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Repin1 | NM_001079901.1 | NP_001073370.2 | ||
| NM_001079902.1 | NP_001073371.2 | |||
| NM_001079903.1 | NP_001073372.2 | |||
| NM_001079904.1 | NP_001073373.2 | |||
| NM_001079905.1 | NP_001073374.1 | |||
| NM_175099.3 | NP_780308.2 | |||
| AK089041.1 | BAC40716.1 | |||
| AK137310.1 | ||||
| AK170797.1 | ||||
| BB870810.1 | ||||
| BC085128.1 | AAH85128.1 | |||
| BC092301.1 | AAH92301.1 | |||
| BM943840.1 | ||||
| BY015371.1 | ||||
| BY145852.1 |
More Information
Set Membership: |
|

Genomic Map