Assay Details
Target Gene Details
Entrez Gene ID: | 791299 |
Gene Name: | predicted gene 10069 |
Gene Aliases: |
ENSMUSG00000059659 |
Location: |
Chr.6:128438757-128503281 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gm10069 | NR_028593.1 | 3 | 897 |
Target Gene Details
Entrez Gene ID: | 11287 |
Gene Name: | pregnancy zone protein |
Gene Aliases: |
A1m, A2m, AI893533, MAM |
Location: |
Chr.6:128483567-128526720 on Build GRCm38 |
Assay Gene Location: | Within Exon 29 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pzp | NM_007376.4 | 29 | 3798 | NP_031402.3 |
| BC057983.1 | 29 | 3798 | AAH57983.1 | |
| BI145862.1 | 2 | 352 | ||
| BI147927.1 | 2 | 256 | ||
| M93264.1 | 29 | 3791 | AAA39508.1 |
More Information
Set Membership: |
|

Genomic Map