Assay Details
Target Gene Details
Entrez Gene ID: | 232585 |
Gene Name: | von Willebrand factor D and EGF domains |
Gene Aliases: |
Gm467 |
Location: |
Chr.6:13185610-13224965 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vwde | NM_001013757.2 | NP_001013779.2 | ||
| BC076621.1 | AAH76621.1 | |||
| BC151200.1 |

Genomic Map