Assay Details
Target Gene Details
Entrez Gene ID: | 22685 |
Gene Name: | zinc finger protein 239 |
Gene Aliases: |
Mok-2, Mok2, Znf239 |
Location: |
Chr.6:117862081-117872766 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp239 | NM_001001792.1 | NP_001001792.1 | ||
| NM_008616.2 | NP_032642.2 | |||
| AK078513.1 | ||||
| AK163713.1 | 1 | 74 | ||
| AK166228.1 | ||||
| BE647041.1 | ||||
| BY128335.1 |
More Information
Set Membership: |
|

Genomic Map