Assay Details
Target Gene Details
Entrez Gene ID: | 56463 |
Gene Name: | staphylococcal nuclease and tudor domain containing 1 |
Gene Aliases: |
AL033314, Tudor-SN |
Location: |
Chr.6:28480315-28935162 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snd1 | NM_019776.2 | 2 | 370 | NP_062750.2 |
| AB021491.1 | 2 | 327 | BAA84944.1 | |
| AK034914.1 | 2 | 318 | ||
| AK088028.1 | 2 | 329 | BAC40105.1 | |
| AK133837.1 | 2 | 376 | BAE21874.1 | |
| AK159837.1 | 2 | 335 | BAE35415.1 | |
| AK161591.1 | 2 | 324 | BAE36479.1 | |
| AK162432.1 | 2 | 311 | BAE36913.1 | |
| AK167576.1 | 2 | 360 | BAE39639.1 | |
| AK204079.1 | ||||
| BB613775.1 | 2 | 370 | ||
| BC007126.1 | 2 | 317 | AAH07126.3 | |
| BM944959.1 |

Genomic Map