Assay Details
Target Gene Details
Entrez Gene ID: | 56463 |
Gene Name: | staphylococcal nuclease and tudor domain containing 1 |
Gene Aliases: |
AL033314, Tudor-SN |
Location: |
Chr.6:28480315-28935162 on Build GRCm38 |
Assay Gene Location: | Within Intron 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Snd1 | NM_019776.2 | NP_062750.2 | ||
| AB021491.1 | BAA84944.1 | |||
| AK088028.1 | BAC40105.1 | |||
| AK133837.1 | BAE21874.1 | |||
| AK159837.1 | BAE35415.1 | |||
| AK161591.1 | BAE36479.1 | |||
| AK162432.1 | BAE36913.1 | |||
| AK167576.1 | BAE39639.1 | |||
| AK181132.1 | ||||
| BC007126.1 | AAH07126.3 |
More Information
Set Membership: |
|

Genomic Map